Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Methyleenchloride ≥99% stabilised, GPR RECTAPUR®

Methyleenchloride ≥99% stabilised, GPR RECTAPUR®

Metal drum, 25 L
Catalog # 25631.464
Health
Supplier:   VWR Chemicals
Methyleenchloride ≥99% stabilised, GPR RECTAPUR®
Catalog # 25631.464
Supplier:  VWR Chemicals
CAS Number:  75-09-2
Methyleenchloride ≥99% stabilised, GPR RECTAPUR®
Methyleenchloride ≥99% stabilised, GPR RECTAPUR®

Metal drum, 25 L

CAS Number:  75-09-2
Price
341,00 €
Log in for availability
1 * 25 L
Product Details & Documents
Synonyms:  Methylene chloride, DCM
  • CAS Number:75-09-2
  • Formula:CH₂Cl₂
  • MDL Number:MFCD00000881
  • Molecular Weight:84.93 g/mol
  • Merck Index:12,06140
  • Boiling Pt:39,8 °C (1013 hPa)
  • Melting Pt:–95 °C
  • Density:1,322 g/cm³ (20 °C)
  • EINECS:200-838-9
  • ADR:6.1,III
  •  :Stabilised with ethanol 0,1 %
  • Storage Temperature:Kamertemperatuur
  • UN:1593
  • REACH:01-2119480404-41
Specifications

Specification Test Results

  • Assay:
    Min. 99 %
  • Acidity:
    Max. 0.0004 meq/g
  • Boiling point:
    39.0 - 40.5 °C
  • Density (20/4):
    1.322 - 1.327
  • Evaporation residue:
    Max. 20 ppm
  • Water:
    Max. 0.02 %

Specifications

  • Verpakking:
    Metal drum
  • VPE:
    25 L
  • Cat. No.:
    25631.464

Specifications