Methyleenchloride ≥99% stabilised, GPR RECTAPUR®
Plastic bottle, fluorinated, 5 L
Leverancier: VWR Chemicals
Methyleenchloride ≥99% stabilised, GPR RECTAPUR®
Catalogus # 25631.362
Leverancier: VWR Chemicals
CAS-nummer: 75-09-2
Ga naar:
Product Details & Documents
Synoniemen: Methylene chloride, DCM
- CAS-nummer:75-09-2
- Formule:CH₂Cl₂
- MDL-nummer:MFCD00000881
- Molecular Weight:84.93 g/mol
- Merck index:12,06140
- Kokend Pt:39,8 °C (1013 hPa)
- Smelten Pt:–95 °C
- Dichtheid:1,322 g/cm³ (20 °C)
- EINECS:200-838-9
- ADR:6.1,III
- :Stabilised with ethanol 0,1 %
- Temperatuur bij opslag:Kamertemperatuur
- UN:1593
- BEREIKEN:01-2119480404-41
Certificaten
Zoeken op lotnummer1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Glass bottle for liquids DIN 45 (ZPACK13415249) 216L Metal drum for liquids UN X (ZPACK10843202) 28L Metal drum for liquids UN X (ZPACK12124128) 30L Metal drum for liquids UN X (ZPACK12124125)
More
Specificaties
Specificatie testresultaten
- Assay:Min. 99 %
- Acidity:Max. 0.0004 meq/g
- Boiling point:39.0 - 40.5 °C
- Density (20/4):1.322 - 1.327
- Evaporation residue:Max. 20 ppm
- Water:Max. 0.02 %
Specificaties
- Verpakking:Plastic bottle, fluorinated
- VPE:5 L



