Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Methyleenchloride ≥99% stabilised, GPR RECTAPUR®

Methyleenchloride ≥99% stabilised, GPR RECTAPUR®

Glass bottle, 1 L
Catalogus # 25631.293
Gezondheid
Leverancier:   VWR Chemicals
Methyleenchloride ≥99% stabilised, GPR RECTAPUR®
Catalogus # 25631.293
Leverancier:  VWR Chemicals
CAS-nummer:  75-09-2
Methyleenchloride ≥99% stabilised, GPR RECTAPUR®
Methyleenchloride ≥99% stabilised, GPR RECTAPUR®

Glass bottle, 1 L

CAS-nummer:  75-09-2
Price
32,30 €
Log in voor beschikbaarheid
1 * 1 L
Product Details & Documents
Synoniemen:  Methylene chloride, DCM
  • CAS-nummer:75-09-2
  • Formule:CH₂Cl₂
  • MDL-nummer:MFCD00000881
  • Molecular Weight:84.93 g/mol
  • Merck index:12,06140
  • Kokend Pt:39,8 °C (1013 hPa)
  • Smelten Pt:–95 °C
  • Dichtheid:1,322 g/cm³ (20 °C)
  • EINECS:200-838-9
  • ADR:6.1,III
  •  :Stabilised with ethanol 0,1 %
  • Temperatuur bij opslag:Kamertemperatuur
  • UN:1593
  • BEREIKEN:01-2119480404-41
Specificaties

Specificatie testresultaten

  • Assay:
    Min. 99 %
  • Acidity:
    Max. 0.0004 meq/g
  • Boiling point:
    39.0 - 40.5 °C
  • Density (20/4):
    1.322 - 1.327
  • Evaporation residue:
    Max. 20 ppm
  • Water:
    Max. 0.02 %

Specificaties

  • Verpakking:
    Glass bottle
  • VPE:
    1 L

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.