Orders LinkContactUsLinkComponent
Racks for cooling circulators
Racks for cooling circulators
Stainless steel rack for 1×in AL 5, 3×in AL 12, 5×in AL 18, AL 25, 21, 14/18 mm, 80 mm high
Catalog # 462-1009
Supplier:   LAUDA
Racks for cooling circulators
Catalog # 462-1009
Supplier:  LAUDA
Racks for cooling circulators
Stainless steel rack for 1×in AL 5, 3×in AL 12, 5×in AL 18, AL 25, 21, 14/18 mm, 80 mm high
Price
308,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Voor buizen Ø:
    14/18 mm
  • Voor buizen:
    80 mm high
  • Aant. gaten:
    21
  • Omschrijving:
    Stainless steel rack for 1×in AL 5, 3×in AL 12, 5×in AL 18, AL 25
  • Seq:
    02