Orders LinkContactUsLinkComponent
Passieve dositubes
Passieve dositubes
Koolmonoxide (1DL), Carbon monoxide, 0,4 - 400 ppm
Catalog # 151-0082
Supplier:   GASTEC
Passieve dositubes
Catalog # 151-0082
Supplier:  GASTEC
Passieve dositubes
Passieve dositubes
Koolmonoxide (1DL), Carbon monoxide, 0,4 - 400 ppm
Price
53,10 €
Log in for availability
1 * 10 ST
Product Details & Documents
The dositubes are used to measure time-weighted average (TWA) gas concentrations over longer periods.
  • Compact, lightweight, and versatile (can be used anywhere)
  • Easy direct measuring and reading
  • No sampling pump necessary
The Dositubes use the principle of the natural diffusion of gas.
A Dositube is a glass tube of a specific inner diameter that is filled with a diffuser and chemical reagent in parallel that enable gas to disperse at a constant rate resulting in a longer discolouration and better demarcation. The passive Dositube can then be positioned in the worker's breathing zone where it can be attached with a holder and collar clip.
Specifications
  • Omschrijving:
    Koolmonoxide (1DL)
  • Parameter:
    Carbon monoxide
  • Meetbereik:
    0,4 - 400 ppm
  • Cat. No.:
    151-0082
  • Meetduur:
    0,5 - 24 h
Accessories
Product Information
  • Catalog #
    Omschrijving
    Parameter
    Meetbereik
    Availability
    Price
  • 151-0090
    Azijnzuur (81D)
    Acetic acid
    0,5 - 100 ppm
    Log in for availability
    1 * 10 ST - 53,10 €
Product Information
  • Catalog #
    Omschrijving
    Availability
    Price
  • 151-0095
    Houder voor Dositube
    Log in for availability
    1 * 3 ST - 39,80 €