Orders LinkContactUsLinkComponent
Passieve dositubes
Passieve dositubes
Methylethylketon (152D), Methyl ethyl ketone, 2 - 600 ppm
Catalogus # 151-0078
Leverancier:   GASTEC
Passieve dositubes
Catalogus # 151-0078
Leverancier:  GASTEC
Passieve dositubes
Passieve dositubes
Methylethylketon (152D), Methyl ethyl ketone, 2 - 600 ppm
Price
53,10 €
Log in voor beschikbaarheid
1 * 10 ST
Product Details & Documents
The dositubes are used to measure time-weighted average (TWA) gas concentrations over longer periods.
  • Compact, lightweight, and versatile (can be used anywhere)
  • Easy direct measuring and reading
  • No sampling pump necessary
The Dositubes use the principle of the natural diffusion of gas.
A Dositube is a glass tube of a specific inner diameter that is filled with a diffuser and chemical reagent in parallel that enable gas to disperse at a constant rate resulting in a longer discolouration and better demarcation. The passive Dositube can then be positioned in the worker's breathing zone where it can be attached with a holder and collar clip.
Specificaties
  • Omschrijving:
    Methylethylketon (152D)
  • Parameter:
    Methyl ethyl ketone
  • Meetbereik:
    2 - 600 ppm
  • Cat. No.:
    151-0078
  • Meetduur:
    1 -10 h
Accessories
Productinformatie
  • Catalogus #
    Parameter
    Meetbereik
    Beschikbaarheid
    Price
  • Azijnzuur (81D)
    Azijnzuur (81D)
    151-0090
    Acetic acid
    0,5 - 100 ppm
    Log in voor beschikbaarheid
    1 * 10 ST - 53,10 €
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Houder voor Dositube
    Houder voor Dositube
    151-0095
    Log in voor beschikbaarheid
    1 * 3 ST - 39,80 €