Orders LinkContactUsLinkComponent
Instrument boxes
Instrument boxes
blauw, 100×215×50 mm
Catalogus # AGEN8036B
Leverancier:   Agencinox
Instrument boxes
Catalogus # AGEN8036B
Leverancier:  Agencinox
Instrument boxes
Instrument boxes
blauw, 100×215×50 mm
Price
68,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Aluminium, anodised.
  • Smooth surfaces for fast, easy cleaning
  • Coloured
  • Good chemical resistance, very good thermal resistance
Specificaties
  • Kleur:
    blauw
  • Seq:
    06
  • b×d×h:
    100×215×50 mm