Spatulas
Spatula, spear-shaped (G), 140 mm
Catalogus # 231-0000
Leverancier: Agencinox
Ga naar:
Product Details & Documents
Stainless steel ¹⁸/₁₀, anti-magnetic, electro-polishing. These universal spatulas are suitable for many applications.
- Zeer goede thermische bestendigheid, autoclaveerbaar
- Grote weerstand tegen chemische stoffen
Specificaties
- Omschrijving:Spatula, spear-shaped (G)
- Lengte:140 mm