Orders LinkContactUsLinkComponent
Kroestangen, robuust model
Kroestangen, robuust model
400 mm, 35 mm
Catalog # BOCH2144
Supplier:   BOCHEM
Kroestangen, robuust model
Catalog # BOCH2144
Supplier:  BOCHEM
Kroestangen, robuust model
Kroestangen, robuust model
400 mm, 35 mm
Price
32,50 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Crucible tongs made from electrolytically polished stainless steel.
Spécifications
  • Lengte:
    400 mm
  • Seq:
    05
  • Spanbereik:
    35 mm