0,00 €
Bestelling invoeren
/
Products
Services
Digital Solutions
Applications
Support
About
Belgium
Orders Link
ContactUsLinkComponent
Tangen
Kroestangen, robuust model
Kroestangen, robuust model
220 mm, 20 mm
BOCH2141
:
BOCHEM
Kroestangen, robuust model
BOCH2141
:
BOCHEM
Kroestangen, robuust model
220 mm, 20 mm
17,30 €
1 * 1 ST
-
+
Crucible tongs made from electrolytically polished stainless steel.
Lengte:
220 mm
Seq:
02
Spanbereik:
20 mm