Orders LinkContactUsLinkComponent
Druppelflessen, WHEATON®
15 - 415, groen
Catalogus # 215-4016
Leverancier:   DWK Life Sciences
Druppelflessen, WHEATON®
Catalogus # 215-4016
Leverancier:  DWK Life Sciences
Druppelflessen, WHEATON®
Druppelflessen, WHEATON®
15 - 415, groen
Price
42,10 €
Log in voor beschikbaarheid
1 * 100 ST
Product Details & Documents
Screw cap made of radiation grade polypropylene.
  • Very good chemical resistance
  • Very good temperature resistance
  • Variety of colors
Specificaties
  • Schroefdraad:
    15 - 415
  • Seq:
    29
  • Kleur dop:
    groen