Push-on caps for sample bottles
Natural PE cap, Ø 30 mm
215-2700
: GRAVIS
- Omschrijving:Natural PE cap, Ø 30 mm
- Seq:05
Reagents for water determination by the Coulometric Titration Karl Fischer method, Hydranal™
#: 34820-500ML
: Honeywell Chemicals
Ijsazijn glacial 99.8-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
#: 20104.243
: VWR Chemicals
Tetrahydrofuraan stabilised, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
#: 1.09731.2500
: Merck
Tetrahydrofuraan ≥99.5% (by anhydrous basis) stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
#: 28551.321
: VWR Chemicals
Xylenol orange triturate 1% (w/w) in potassium nitrate Reag. Ph. Eur. 1096301
#: 87963.150
: VWR Chemicals