Trypsinizing flasks, WHEATON®
700 ml, Schroefdop voor bovenzijde 51-400, Schroefdop 20-415
N° de catalogue WHEA355807
Fournisseur: DWK Life Sciences
Aller à :
Product Details & Documents
Borosilicate glass, clear.
- With pour out and caps
- Fluted design improves mixing
- Deep skirted screw caps
These flasks feature deep-fluted sides to promote mixing. A dam below the sidearm retains untrypsinized tissue when decanting. A deep skirted screw cap on the sidearm prevents contamination of flask contents. The sidearm allows easier decanting of contents while retaining large particles behind the dam. Sidearm caps are 20-415 size. Caps have white styrene-butadiene rubber liners.
Spécifications
- Capaciteit:700 ml
- Omschrijving:Schroefdop voor bovenzijde 51-400
- Hals:Schroefdop 20-415