Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Maatkolven

Maatkolven

2000 ml, 370 mm, 0,600 ± ml
Catalog # 612-0094
Supplier:   SciLabware
Maatkolven
Catalog # 612-0094
Supplier:  SciLabware
Maatkolven
Maatkolven

2000 ml, 370 mm, 0,600 ± ml

Price
53,70 €
Log in for availability
1 * 1 ST
Product Details & Documents

MBL®, borosilicate glass, with smooth neck.

  • Inscriptions in blue enamel
  • Conformity certified, with batch number

ISO 1042

Certifications: ISO 1042

Specifications
  • Capaciteit:
    2000 ml
  • Hoogte:
    370 mm
  • Tolerantie:
    0,600 ± ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.