Orders LinkContactUsLinkComponent
Kranen
Kranen
Elbow hose, On table
Catalogus # 132-2704
Leverancier:   EQUIP LABO
Kranen
Catalogus # 132-2704
Leverancier:  EQUIP LABO
Kranen
Kranen
Elbow hose, On table
Price
32,90 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
The taps are of 3 types:
  • Taps for hard and demineralised water
  • Taps for combustible gases
  • Taps for air, compressed gas and vacuum
Straight hose length 80 cm, exterior stainless steel braid, interior neoprene sheath. Suitable for taps on the table-top (for cold water, hot water, compressed air). Outlet by 15/21 female nut for connection on 15/21 (¹/₂") male standby gas feed.
Elbow hose with same characteristics as straight hoses but adapted to plumbing fittings on splash back (for cold water, hot water, compressed air).
 : Different sockets (individual and in a row) are available on request.
 : * All of these valves are available in a counter-top version.
 : Installatie, IQOQ, onderhoud of reparatie nodig? Mail naar [email protected]
Specificaties
  • Omschrijving:
    Elbow hose, On table