Kranen
The taps are of 3 types:
- Taps for hard and demineralised water
- Taps for combustible gases
- Taps for air, compressed gas and vacuum
Straight hose length 80 cm, exterior stainless steel braid, interior neoprene sheath. Suitable for taps on the table-top (for cold water, hot water, compressed air). Outlet by 15/21 female nut for connection on 15/21 (¹/₂") male standby gas feed.
Elbow hose with same characteristics as straight hoses but adapted to plumbing fittings on splash back (for cold water, hot water, compressed air).
Ordering information: Different sockets (individual and in a row) are available on request.
Caution: * All of these valves are available in a counter-top version.
Services: Installatie, IQOQ, onderhoud of reparatie nodig? Mail naar [email protected]
- Omschrijving:Combustible gas tap, On backsplash