Nessler reagent, tablets
DPD 4, 250
Catalogus # LOVI511261BT
Leverancier: Lovibond
Ga naar:
Product Details & Documents
Synoniemen: Potassium mercuric Iodide, Potassium tetraiodomercurate, Nota 1, Mercury (II) potassium iodide, Potassium tetraiodomercurate – Potassium hydroxide solution
- CAS-nummer:7783-33-7
- MDL-nummer:MFCD00049656
Documents
Certificaten
Zoeken op lotnummerSpecificaties
Specificaties
- VPE:250
- Info VPE:DPD 4
Specificaties
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



