O-Ring seals for flat flanges
Vlakke flenzen DN 100
Catalogus # 201-0152
Leverancier: Lenz Laborglas
Ga naar:
Product Details & Documents
Silicone or Silicone PTFE-coated.
- Excellent chemical resistance
- Excellent temperature resistance
Seals for flat flanges LF 60-200.
Specificaties
- Voor:Vlakke flenzen DN 100
- Seq:02