Orders LinkContactUsLinkComponent
Distilling receivers, Bernauer
Distilling receivers, Bernauer
29/32 NS
Catalog # LENZ05288104
Supplier:   Lenz Laborglas
Distilling receivers, Bernauer
Catalog # LENZ05288104
Supplier:  Lenz Laborglas
Distilling receivers, Bernauer
Distilling receivers, Bernauer
29/32 NS
Price
99,20 €
Log in for availability
1 * 1 ST
Product Details & Documents
DURAN®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Distilling receivers acc. to Bernauer (modified), with three cones NS 14/23, vacuum connection with glass olive. With flasks 100 ml capacity or without flasks available.
Documents
Specifications
  • Huls:
    29/32 NS