Orders LinkContactUsLinkComponent
Apparatus - Antonacopoulos
Spare spiral condenser with socket, - NS, 29/32 NS
Catalog # LENZ05434005
Supplier:   Lenz Laborglas
Apparatus - Antonacopoulos
Catalog # LENZ05434005
Supplier:  Lenz Laborglas
Apparatus - Antonacopoulos
Spare spiral condenser with socket, - NS, 29/32 NS
Price
108,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
DURAN®, Borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Specifications
  • Afmetingen kern:
    - NS
  • Huls:
    29/32 NS
  • Omschrijving:
    Spare spiral condenser with socket
  • Seq:
    05