Apparatus - Antonacopoulos
Spare adapter, 70/50 NS, 29/32 NS
LENZ05434002
DURAN®, Borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
- Afmetingen kern:70/50 NS
- Huls:29/32 NS
- Omschrijving:Spare adapter
- Seq:03