Orders LinkContactUsLinkComponent
Apparatus - Antonacopoulos
Apparatus - Antonacopoulos
Spare flask, 2000 ml capacity, 70/50 NS, - NS
Catalogus # LENZ05434001
Leverancier:   Lenz Laborglas
Apparatus - Antonacopoulos
Catalogus # LENZ05434001
Leverancier:  Lenz Laborglas
Apparatus - Antonacopoulos
Spare flask, 2000 ml capacity, 70/50 NS, - NS
Price
311,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
DURAN®, Borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Documents
Specificaties
  • Afmetingen kern:
    70/50 NS
  • Huls:
    - NS
  • Omschrijving:
    Spare flask, 2000 ml capacity
  • Seq:
    02