Orders LinkContactUsLinkComponent
Nessler-cilinders
Nessler-cilinders
Nessler cylinders, 50 ml, low form, Ø×H: 26×175, 50 ml division
Catalog # 214-0180
Nessler-cilinders
Catalog # 214-0180
Nessler-cilinders
Nessler cylinders, 50 ml, low form, Ø×H: 26×175, 50 ml division
Price
52,20 €
Log in for availability
1 * 1 ST
Product Details & Documents
  • Optisch gepolijst, vlakke voet
  • Met schenktuit
Specifications
  • Omschrijving:
    Nessler cylinders, 50 ml, low form, Ø×H: 26×175, 50 ml division
  • Seq:
    04