Handheld refractometers
0 - 20 °Brix, 0,1 °Brix
Catalogus # 635-0309
Leverancier: EUROMEX
Ga naar:
Product Details & Documents
Single dual or triple scales, analogue, handheld refractometers.
- Automated temperature compensation (ATC)
- Calibrated at 20 °C except for RF.5190
- Requires only a few drops of liquid
: Installatie, IQOQ, onderhoud of reparatie nodig? Mail naar [email protected]
Specificaties
- Meetbereik:0 - 20 °Brix
- Seq:01
- Precisie:0,1 °Brix