Orders LinkContactUsLinkComponent
Cary source lamps
Cary source lamps
Visible source lamp, for Cary 100/300
Catalogus # VARI5610021700
Leverancier:   VARIAN
Cary source lamps
Catalogus # VARI5610021700
Leverancier:  VARIAN
Cary source lamps
Cary source lamps
Visible source lamp, for Cary 100/300
Price
62,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Source lamps for Cary spectrophotometers are fully modular and interchangeable. A range of UV, xenon and visible lamps are available.
  • Fully modular and compatible with Cary spectrophotometers
The UV, xenon and visible lamps are fully modular, allowing you to select and change between light sources according to your experimental requirements.
Specificaties
  • Omschrijving:
    Visible source lamp, for Cary 100/300
  • Seq:
    03
Accessories
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Loom, deuterium lamp extension, required when installing deuterium lamp mm a Cary ¹/₃/100/300 series UV-Vis
    Loom, deuterium lamp extension, required when installing deuterium lamp mm a Cary ¹/₃/100/300 series UV-Vis
    HEWLG9820-60000
    Log in voor beschikbaarheid
    1 * 1 ST - 37,00 €