Orders LinkContactUsLinkComponent
Cary source lamps
Cary source lamps
Lamp, visible, quartz-iodide (QI), for Cary 4000/5000/6000i
Catalogus # VARI5610013900
Leverancier:   VARIAN
Cary source lamps
Catalogus # VARI5610013900
Leverancier:  VARIAN
Cary source lamps
Cary source lamps
Lamp, visible, quartz-iodide (QI), for Cary 4000/5000/6000i
Price
57,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Source lamps for Cary spectrophotometers are fully modular and interchangeable. A range of UV, xenon and visible lamps are available.
  • Fully modular and compatible with Cary spectrophotometers
The UV, xenon and visible lamps are fully modular, allowing you to select and change between light sources according to your experimental requirements.
Specificaties
  • Omschrijving:
    Lamp, visible, quartz-iodide (QI), for Cary 4000/5000/6000i
  • Seq:
    02
Accessories
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Loom, deuterium lamp extension, required when installing deuterium lamp mm a Cary ¹/₃/100/300 series UV-Vis
    Loom, deuterium lamp extension, required when installing deuterium lamp mm a Cary ¹/₃/100/300 series UV-Vis
    HEWLG9820-60000
    Log in voor beschikbaarheid
    1 * 1 ST - 37,00 €