Capillary columns, cyclodex-B
on 7" cage, Capillair, Cyclodex-B, 0,25 µm, 0,25 mm, 60 m
HEWL112-2562
: Agilent
Cyclodex-B capillary columns provides excellent peak shape.
- 10,5% β-cyclodextrin in DB-1701
- Chiral separations without chiral-specific derivatisation
- Broad range of resolving potential
- Kolomtype:Capillair
- Filmdikte:0,25 µm
- Ø inw.:0,25 mm
- Omschrijving:on 7" cage
- Lengte:60 m
- Seq:06
- Fase:Cyclodex-B