Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Dimethylsulfoxide

Dimethylsulfoxide

Glass bottle, 500 ml
Catalog # BMOL54686-2
Dimethylsulfoxide
Catalog # BMOL54686-2
CAS Number:  67-68-5
Dimethylsulfoxide
Dimethylsulfoxide

Glass bottle, 500 ml

CAS Number:  67-68-5
Price
Inquire For Price
Log in for availability
1 * 500 mL
Product Details & Documents
Synonyms:  Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO
  • CAS Number:67-68-5
  • Formula:(CH₃)₂SO
  • MDL Number:MFCD00002089
  • Molecular Weight:78.14 g/mol
  • Merck Index:13,03285
  • Boiling Pt:189 °C (1013 hPa)
  • Melting Pt:18,5 °C
  • Density:1,101 g/cm³ (20 °C)
  • Flash Pt:87 °C
  • Storage Temperature:Kamertemperatuur
  • EINECS:200-664-3
Specifications

Specifications

  • Verpakking:
    Glass bottle
  • VPE:
    500 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.