Natriumdodecylsulfaat
Glass bottle for solids, 500 g
BMOL51430-2
: Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate
- :151-21-3
- :CH₃(CH₂)₁₁OSO₃Na
- :MFCD00036175
- :288.38 g/mol
- :14,08636
- :204…207 °C
- :1,1 g/cm³ (20 °C)
- :>100 °C
- :Kamertemperatuur
- :205-788-1
- :1325
- :4.1,II
- Verpakking:Glass bottle for solids
- VPE:500 g