Boorzuur
5 kg
Catalogus # BMOL50765-3
Leverancier: BIOMOL RESEARCH LABORATORIES
Ga naar:
Product Details & Documents
Synoniemen: Boron trihydroxide, Boron hydroxide, Boric acid, Boracic acid
- CAS-nummer:10043-35-3
- Formule:B(OH)₃
- MDL-nummer:MFCD00011337
- Molecular Weight:61.83 g/mol
- Merck index:13,01326
- Kokend Pt:185 °C (1013 hPa)
- Smelten Pt:160 °C
- Dichtheid:1,44 g/cm³ (25 °C)
- Temperatuur bij opslag:Kamertemperatuur
- EINECS:233-139-2
- BEREIKEN:01-2119486683-25
Specificaties
Specificaties
- VPE:5 kg
Specificaties
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



