Boorzuur
Glass bottle for solids, 1 kg
BMOL50765-1
: Boron trihydroxide, Boron hydroxide, Boric acid, Boracic acid
- :10043-35-3
- :B(OH)₃
- :MFCD00011337
- :61.83 g/mol
- :13,01326
- :185 °C (1013 hPa)
- :160 °C
- :1,44 g/cm³ (25 °C)
- :Kamertemperatuur
- :233-139-2
- :01-2119486683-25
- Verpakking:Glass bottle for solids
- VPE:1 kg