Pepstatin A
Glass bottle for solids, 25 mg
Catalog # BMOL17640-1
Supplier: BIOMOL RESEARCH LABORATORIES
Jump to:
Product Details & Documents
Specifications
Specifications
- Verpakking:Glass bottle for solids
- VPE:25 mg
- Cat. No.:BMOL17640-1
- Molecular formula:C₃₄H₆₃N₅O₉
- Protein/peptide name:Pepstatin A
- MDL No.:MFCD00060740
- CAS No.:26305-03-3
- Opslag condities:Refrigerator



