Temperature control jackets for viscosity cups
Temperature control jacket with tripod for DIN and ASTM cups
Catalogus # 769-1131
Leverancier: Industrial Physics
Ga naar:
Product Details & Documents
The temperature control jacket for viscosity cups is a double jacketed casing of anodised aluminium with built-in spirit level to bring the viscosity beaker to the required temperature with cooled and heated liquids.
- Adjustable feet for easy leveling
- Built-in spirit level to set jacket exactly horizontal
- Glass plate to prevent dripping before measurement
- Elbow-type quick release couplings
- Titanium anodised aluminium
Levering: Supplied with jacket with built-in spirit level, glass plate and tube connection.
Diensten: Installatie, IQOQ, onderhoud of reparatie nodig? Mail naar [email protected]
Documents
TQC Company Video
Video 's
Specificaties
- Omschrijving:Temperature control jacket with tripod for DIN and ASTM cups
- Seq:01