Milk butyrometer
Butyrometer for milk, 0 - 9% : 0,1
Catalogus # 614-2006
Leverancier: GERBER FUNKE
Ga naar:
Product Details & Documents
Borosilicate glass.
- With or without stopper
Available with a declaration of conformity per batch and type (in German only).
Specificaties
- Omschrijving:Butyrometer for milk, 0 - 9% : 0,1
- Seq:12
Accessories
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Stopper, rubber, conical, for all measuring method butyrometersStopper, rubber, conical, for all measuring method butyrometers614-2028Log in voor beschikbaarheid1 * 100 ST - 48,20 €