Orders LinkContactUsLinkComponent
Analogue stopwatches, split-second addition timer
Analoge stopwatch, pin hendel, 7 jewels, 1/5 seconden, 30 minuten, 55 mm
Catalogus # 609-1129
Leverancier:   HANHART
Analogue stopwatches, split-second addition timer
Catalogus # 609-1129
Leverancier:  HANHART
Analogue stopwatches, split-second addition timer
Analogue stopwatches, split-second addition timer
Analoge stopwatch, pin hendel, 7 jewels, 1/5 seconden, 30 minuten, 55 mm
Price
742,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Addition timer for time measurement with interrupt option, also drag indicator for split-time measurement.
  • Chrome plated brass casing, diamond cut
  • Lever, 7 jewels
  • Shock resistant, additional dust and water protection
Services: Voor installatie, IQOQ, erkende kalibratie onder ISO17025, onderhoud en reparatie mail naar [email protected]
Specificaties
  • Ø:
    55 mm
  • Omschrijving:
    Analoge stopwatch, pin hendel, 7 jewels, 1/5 seconden, 30 minuten