Dop
48 mm
Catalogus # RIXI8-1080-48_U
Leverancier: BERLIN PACKAGING RIXIUS GMBH
Ga naar:
Product Details & Documents
Metal, white.
- Suitable for screw cap jars 216-3620 to 216-3624
- Good leakproof properties
- Convenient pack sizes
Documents
RIXI8-1080-43_TDS_EN
Technische fiche
Specificaties
- Schroefdraad:48 mm
- Seq:02