Orders LinkContactUsLinkComponent
Monsternameflessen
Monsternameflessen
9,80 ml, 18×65 mm, Ja
Catalogus # 215-8345
Leverancier:   MUELLER KREMPEL
Monsternameflessen
Catalogus # 215-8345
Leverancier:  MUELLER KREMPEL
Monsternameflessen
Monsternameflessen
9,80 ml, 18×65 mm, Ja
Price
258,00 €
Log in voor beschikbaarheid
1 * 266 ST
Product Details & Documents
Vials made from AR glass, clear with black screw cap. Ideal for taking, storing and transporting samples.
  • Thin-walled, cylindrical
  • Tightly sealing screw cap
  • Designed for use in applications that do not require resistance to temperature
Specificaties
  • Inclusief dop:
    Ja
  • Ø×h:
    18×65 mm
  • Capaciteit:
    9,80 ml