Monsternameflessen
7,50 ml, 16×60 mm, Ja
Catalog # 215-8344
Supplier: MUELLER KREMPEL
Jump to:
Product Details & Documents
Vials made from AR glass, clear with black screw cap. Ideal for taking, storing and transporting samples.
- Thin-walled, cylindrical
- Tightly sealing screw cap
- Designed for use in applications that do not require resistance to temperature
Specifications
- Inclusief dop:Ja
- Ø×h:16×60 mm
- Capaciteit:7,50 ml