Orders LinkContactUsLinkComponent
pSTBlue-1 AccepTor™ vector kit
pSTBlue-1 AccepTor™ vector kit
pSTBlue-1 AccepTor™ Vector Kit, 40 Tests
Catalogus # 70595-4
pSTBlue-1 AccepTor™ vector kit
Catalogus # 70595-4
CAS-nummer:  
pSTBlue-1 AccepTor™ vector kit
pSTBlue-1 AccepTor™ Vector Kit, 40 Tests
CAS-nummer:  
Price
821,00 €
Log in voor beschikbaarheid
1 * 40 Tests
Product Details & Documents
The pSTBlue-1 AccepTor™ vector kits are designed to simplify the PCR cloning process by using non-proofreading thermostable DNA polymerases (eg. KOD XL polymerase and native and recombinant Taq polymerases), which leave single 3'-dA overhangs on the reaction products. The AccepTor™ vectors enable direct ligation by virtue of single 3'-dU overhangs that anneal efficiently with 3'-dA extensions on PCR products. The dU residues are converted to dT residues in vivo following transformation.
Levering: AccepTor™ vectors are supplied in a ready-to-ligate format.
Specificaties
Specificaties
  • VPE:
    40 Tests
  • Omschrijving:
    pSTBlue-1 AccepTor™ Vector Kit
Specificaties
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.