PH-combinatie-elektroden
Sensolyt® ECA pH single rod measuring chain**, Sensolyt® ECA, for installation into the SensoLyt® fitting, 2 - 12, Normal contaminated wasterwater
Catalogus # 708-0057
Leverancier: WTW ONLINE
Ga naar:
Product Details & Documents
The SensoLyt® measuring chains with arming are easy to install and boast flexible measuring and application areas. Eletrodes without arming can be connected directly to a pH monitor of the EcoLine® or QuadroLine® series. Depending on the application, they can be installed into a flow vessel.
- Ideal for measurements in water and drinking water
- Also for high pressure and temperature conditions
- Easy to maintain by easy measuring chain replacement
Specificaties
- Voor:Normal contaminated wasterwater
- Type:Sensolyt® ECA, for installation into the SensoLyt® fitting
- Omschrijving:Sensolyt® ECA pH single rod measuring chain**
- Seq:35
- pH:2 - 12
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.