Spider evaporators
Spider with cylindric flasks, SJ24/40, glass, incl. 6 cylindric flasks, SJ14.5/23, 20 ml, For simultaneous distillation from 6×20 ml cylindrical flasks with 24/40 NS Joint
Catalogus # BUCH011576
Ga naar:
Discontinued
Product Details & Documents
Spider evaporators are ideal when users need to concentrate several samples under the same conditions.
- Models for distillation with evaporating or cylindrical flasks available
- Parallel distillation saves time
Specificaties
- Omschrijving:Spider with cylindric flasks, SJ24/40, glass, incl. 6 cylindric flasks, SJ14.5/23, 20 ml
- Type:For simultaneous distillation from 6×20 ml cylindrical flasks with 24/40 NS Joint
- Cat. No.:BUCH011576