Orders LinkContactUsLinkComponent
Multiple adapters
29/32 NS, 29/32 NS
Catalogus # 201-2217
Leverancier:   Lenz Laborglas
Multiple adapters
Catalogus # 201-2217
Leverancier:  Lenz Laborglas
Multiple adapters
Multiple adapters
29/32 NS, 29/32 NS
Price
42,90 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
DURAN®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With one standard ground NS14/23 or NS 29/32 socket and two standard ground NS 14/23 or NS 29/32 cons. Side neck descending angle of 75°.
Specificaties
  • Afmetingen kern:
    29/32 NS
  • Huls:
    29/32 NS