Orders LinkContactUsLinkComponent
Adapter, stopper type, DURAN®
Adapter, stopper type, DURAN®
stopper, solid, 24/29 NS, 19/26 NS
Catalogus # SCOT286576709
Leverancier:   DWK Life Sciences
Adapter, stopper type, DURAN®
Catalogus # SCOT286576709
Leverancier:  DWK Life Sciences
Adapter, stopper type, DURAN®
Adapter, stopper type, DURAN®
stopper, solid, 24/29 NS, 19/26 NS
Price
435,00 €
Log in voor beschikbaarheid
1 * 10 ST
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Adapters, stopper type, solid, socket and cone.
Specificaties
  • Afmetingen kern:
    24/29 NS
  • Huls:
    19/26 NS
  • Type:
    stopper, solid