Orders LinkContactUsLinkComponent
Electrolyte solutions, aqueous
Electrolyte solution, L 2214 2 mol/l KNO3 + 0.001 mol/l KCl, for Ag combination electrodes, thickened, 250 ml in DURAN® glass bottle, 250 mL
Catalog # SCOI285136923
Supplier:   SI Analytics
Electrolyte solutions, aqueous
Catalog # SCOI285136923
Supplier:  SI Analytics
CAS Number:  
Electrolyte solutions, aqueous
Electrolyte solutions, aqueous
Electrolyte solution, L 2214 2 mol/l KNO3 + 0.001 mol/l KCl, for Ag combination electrodes, thickened, 250 ml in DURAN® glass bottle, 250 mL
CAS Number:  
Price
52,20 €
Log in for availability
1 * 250 mL
Product Details & Documents
Electrolyte aqueous solutions are used for reference electrodes, as electrolyte bridges and for storage.
Specifications
Specifications
  • VPE:
    250 mL
  • Omschrijving:
    Electrolyte solution, L 2214 2 mol/l KNO3 + 0.001 mol/l KCl, for Ag combination electrodes, thickened, 250 ml in DURAN® glass bottle
Specifications