Filterkroezen
50 ml, 1, 40 mm
Catalogus # BRND458731
Leverancier: Brand
Ga naar:
Product Details & Documents
These crucibles are manufactured from borosilicate glass 3.3.
Specificaties
- Ø plaat:40 mm
- Porositeit:1
- Capaciteit:50 ml