Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
 

Filterkroezen

30 ml, 2, 30 mm
Catalog # BRND458022
Supplier:   Brand
Filterkroezen
Catalog # BRND458022
Supplier:  Brand
Filterkroezen
Filterkroezen

30 ml, 2, 30 mm

Price
295,00 €
Log in for availability
1 * 10 ST
Product Details & Documents

These crucibles are manufactured from borosilicate glass 3.3.

Specifications
  • Ø plaat:
    30 mm
  • Porositeit:
    2
  • Capaciteit:
    30 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.