Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
 

L(+)-Ascorbic acid, BAKER, Laboratory Reagent, J.T.Baker®

Glass bottle for solids, 100 g
Catalogus # 1914.0100
Leverancier:   Avantor
L(+)-Ascorbic acid, BAKER, Laboratory Reagent, J.T.Baker®
Catalogus # 1914.0100
Leverancier:  Avantor
CAS-nummer:  50-81-7
L(+)-Ascorbic acid, BAKER, Laboratory Reagent, J.T.Baker®
L(+)-Ascorbic acid, BAKER, Laboratory Reagent, J.T.Baker®

Glass bottle for solids, 100 g

CAS-nummer:  50-81-7
Price
51,52 €
Log in voor beschikbaarheid
1 * 100 g
Product Details & Documents
Synoniemen:  Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
  • CAS-nummer:50-81-7
  • Formule:C₆H₈O₆
  • MDL-nummer:MFCD00064328
  • Molecular Weight:176.13 g/mol
  • Merck index:13,00837
  • Kokend Pt:553 °C (1013 hPa)
  • Smelten Pt:190…192 °C
  • Dichtheid:1,694 g/cm³ (20 °C)
  • Temperatuur bij opslag:Kamertemperatuur
  • EINECS:200-066-2
  • BEREIKEN:exempt
Specificaties

Specificatie testresultaten

  • Assay:
    99.0-100.5 %
  • Appearance of solution:
    passes test
  • Copper (Cu):
    max. 5 ppm
  • Heavy Metals (as Pb):
    max. 10 ppm
  • Identification:
    passes test
  • Iron (Fe):
    max. 2 ppm
  • Oxalic acid:
    passes test
  • Specific Optical Rotation:
    +20.5 - +21.5 °
  • Sulfated Ash :
    max. 0,10 %

Specificaties

  • Verpakking:
    Glass bottle for solids
  • VPE:
    100 g

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.