0,00 €
Bestelling invoeren
/
Products
Services
Digital Solutions
Applications
Support
About
Belgium
Orders Link
ContactUsLinkComponent
Scheppen
Weighing scoops
Weighing scoops
70 mm, 32 mm
BOCH12810
:
BOCHEM
Weighing scoops
BOCH12810
:
BOCHEM
Weighing scoops
70 mm, 32 mm
6,90 €
1 * 1 ST
-
+
Aluminium.
Breedte:
32 mm
Lengte:
70 mm
Seq:
01
Double spatulas, BOLA
#: BOHLH915-04
: Bohlender