Toebehoren
Door with window 470×290 mm, interior lighting (30W) 400V 3N ~ 60Hz, ED400
Catalogus # BIND8012-0269
Leverancier: Binder
Ga naar:
Product Details & Documents
Specificaties
- Omschrijving:Door with window 470×290 mm, interior lighting (30W) 400V 3N ~ 60Hz
- Seq:31
- Voor:ED400
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



