Orders LinkContactUsLinkComponent
Pycnometers, unadjusted
Pycnometers, unadjusted
25 ml
Catalogus # 613-2176
Leverancier:   witeg Labortechnik
Pycnometers, unadjusted
Catalogus # 613-2176
Leverancier:  witeg Labortechnik
Pycnometers, unadjusted
Pycnometers, unadjusted
25 ml
Price
22,80 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Pycnometer, made of borosilicate glass 3.3, Gay-Lussac type.
  • With ground-in capillary stopper
  • Nominal capacity printed on the bottom
  • Very good chemical and temperature resistance
Certificeringen: DIN 12797
Specificaties
  • Volume (nom.):
    25 ml